missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Harmol 98.0+%, TCI America™
$233.00 - $922.71
Chemical Identifiers
| CAS | 487-03-6 |
|---|---|
| Molecular Formula | C12H10N2O |
| Molecular Weight (g/mol) | 198.225 |
| MDL Number | MFCD00834164 |
| InChI Key | LBBJNGFCXDOYMQ-UHFFFAOYSA-N |
| Synonym | 7-Hydroxy-1-methyl-9H-pyrido[3,4-b]indole, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol |
| PubChem CID | 68094 |
| IUPAC Name | 1-methyl-2,9-dihydropyrido[3,4-b]indol-7-one |
| SMILES | CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H1360200MG
|
TCI America
H1360200MG |
200 mg |
Each for $233.00
|
|
|||||
|
H13601G
|
TCI America
H13601G |
1 g |
Each for $922.71
|
|
|||||
Chemical Identifiers
| 487-03-6 | |
| 198.225 | |
| LBBJNGFCXDOYMQ-UHFFFAOYSA-N | |
| 68094 | |
| CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1 |
| C12H10N2O | |
| MFCD00834164 | |
| 7-Hydroxy-1-methyl-9H-pyrido[3,4-b]indole, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol | |
| 1-methyl-2,9-dihydropyrido[3,4-b]indol-7-one |
Specifications
| 487-03-6 | |
| Yellow | |
| MFCD00834164 | |
| 7-Hydroxy-1-methyl-9H-pyrido[3,4-b]indole, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol | |
| CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1 | |
| 198.225 | |
| 198.23 | |
| Crystalline Powder |
| 323°C | |
| C12H10N2O | |
| 200 mg | |
| LBBJNGFCXDOYMQ-UHFFFAOYSA-N | |
| 1-methyl-2,9-dihydropyrido[3,4-b]indol-7-one | |
| 68094 | |
| ≥98.0% (T) | |
| Harmol |
Safety and Handling
TSCA : No