missing translation for 'onlineSavingsMsg'
Learn More

Harmol hydrochloride dihydrate, 98%

Catalog No. p-7042805
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAJ6382703 1 g
AAJ6382706 5 g
2 options

Catalog No. AAJ6382703

Supplier: Thermo Scientific Chemicals J6382703

Only null left

Induces apoptosis by Caspase-8 activation | CAS: 149022-16-2 | C12H15ClN2O3 | 270.713 g/mol

Harmol hydrochloride dihydrate is used to induces apoptosis by Caspase-8 activation, CNS stimulant, may act through NMDA receptors, and antitumor agent. Harmol has been shown to inhibit haspin kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Harmol hydrochloride dihydrate is used to induces apoptosis by Caspase-8 activation, CNS stimulant, may act through NMDA receptors, and antitumor agent. Harmol has been shown to inhibit haspin kinase.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 149022-16-2
Molecular Formula C12H15ClN2O3
Molecular Weight (g/mol) 270.713
MDL Number MFCD00150058
InChI Key OITPZRQHDRVLMO-UHFFFAOYSA-N
Synonym harmol hydrochloride dihydrate, 1-methyl-9h-pyrido 3,4-b indol-7-ol hydrochloride dihydrate, harmol hydrochloride hydrate, harmol dihydrate hydrochloride, c12h10n2o.hcl.2h2o, 1-methyl-2,9-dihydropyrido 3,4-b indol-7-one dihydrate hydrochloride
PubChem CID 2723830
IUPAC Name 1-methyl-2,9-dihydropyrido[3,4-b]indol-7-one;dihydrate;hydrochloride
SMILES CC1=C2C(=C3C=CC(=O)C=C3N2)C=CN1.O.O.Cl

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 270.72 (234.69 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Harmol hydrochloride dihydrate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only