missing translation for 'onlineSavingsMsg'
Learn More

Harmine 98.0+%, TCI America™

Catalog No. H00011G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H00011G Supplier TCI America Supplier No. H00011G
Only null left

Chemical Identifiers

CAS 442-51-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004958
InChI Key BXNJHAXVSOCGBA-UHFFFAOYSA-N
Synonym harmine, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, banisterine, leucoharmine, telepathine, yageine, yajeine, 7-methoxy-1-methyl-9h-beta-carboline, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, banisterin
PubChem CID 5280953
ChEBI CHEBI:28121
IUPAC Name 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
SMILES COC1=CC2=C(C=C1)C1=C(N2)C(C)=NC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 212.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Harmine
RTECSNumber UV0175000
TSCA No