missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hardy Diagnostics™ Mycoplasma Agar
Description
Packaged in sleeves of 10 plates in a breathable celluose bag that prevents build-up of condensation and excess moisture
- Mycoplasma bovis
- Prepared media
- Raised rings
- Stacking ability
- Anti-sliding
Certifications
ISO 13485
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | 2°C to 8°C |
| Form | Solid |
| Product Type | Agar |
| Formulation | Noble Agar 15g/L, Cefoperazone 0.07g/L |
| For Use With (Application) | Detection of Mycoplasma species associated with bovine mastitis |
| Packaging Type | Plate |