missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific Supplies Hardware Assortment Kit
Description
- Designed for use by two to four students
- Shipped in a reusable storage box
Order Info
For complete kit contents, please call your Fisher Scientific™ Representative.
Specifications
Specifications
| Grade | 6 to 12 |
| For Use With (Application) | For use in general science, physical science, or chemistry laboratory |
| Includes | A variety of hardware items commonly used in a general science, physical science or chemistry laboratory |
| Class Size | 2 to 4 Students |
| Product Type | Hardware Assortment Kit |