missing translation for 'onlineSavingsMsg'
Learn More

Hardness Titrating Solution, 1 mL = 1 mg CaCO3, Ricca Chemical

For Water Hardness Testing

Supplier:  Ricca Chemical Company R34790004A

Catalog No. R34790004A


Only null left

Description

Description

Specifications

Specifications

Clear colorless
4 L
Natural Poly Bottle
Hardness Titrating Solution
∼ 10
Liquid
4 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

SDS
Documents

Documents

Promotions

Promotions