Learn More
John M. Maris Company Hard Rubber Spatulas
Nonsparking, wide, flexible blade
Supplier: John M. Maris Company 108
Description
Nonsparking, wide, flexible blade
- Integral handle
- Can be used with materials that affect metals adversely
- Washable
- Sterilizable
Chemical Identifiers
| Sterile | |
| Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses | |
| Integral |
| Spatula | |
| Hard Rubber |
Specifications
| Sterile | |
| Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses | |
| 13 in. | |
| 1.37 in. | |
| Hard Rubber | |
| 33 cm |
| Spatula | |
| 8 in. | |
| 20.3 cm | |
| 3.5 cm | |
| Integral |