Learn More
John M. Maris Company Hard Rubber Spatulas
Nonsparking, wide, flexible blade
Supplier:  John M. Maris Company 106/DUP1
Encompass
 View more versions of this product
Catalog No. 14355B
Only null left
Description
Description
Nonsparking, wide, flexible blade
  • Integral handle
  • Can be used with materials that affect metals adversely
  • Washable
  • Sterilizable
Specifications
Chemical Identifiers
Sterile
Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses
Integral
Spatula
Hard Rubber
Specifications
Sterile
Designed to used with materials that affect Metals adversely, for mixing, weighing, cleaning, blending and many other uses
10.25 in.
1.14 in.
Hard Rubber
26 cm
Spatula
6 in.
15.2 cm
2.9 cm
Integral
Product Suggestions
Product Suggestions