missing translation for 'onlineSavingsMsg'
Learn More

Pyramex™ SL Series Hard Hats, 4-point Ratchet

Catalog No. 19161143
Change view
Click to view available options
Color:
Blue
Gray
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19161143 Gray
19161141 Blue
19161137 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19161143 Supplier Pyramex™ Supplier No. HP14112 GRAY

Hard Hats exceed ANSI Z89.1, Type 1, Class C, G, and E for industrial head protection; CSA Z94.1-15.

  • Low profile design
  • Tested and marked for reverse donning
  • Shell constructed from High Density Polyethylene materials
  • Rain trough on sides and back of helmet channel moisture away
  • Soft brow pad is replaceable. Replaceable suspensions and headbands also available
  • Snap Lock suspension features 'tuck away' adjustments for all day, hassle free wear
  • Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat
  • Universal accessory slots fit cap mounted ear muffs and accessories. Hats also accommodate chin straps
  • Testing: Impact, Penetration, Flammability, Electrical Insulation, Marking & Humidity

Specifications

Certifications/Compliance ANSI Z89.1
Color Gray
For Use With (Equipment) Earmuffs, Faceshield
For Use With (Application) Construction
Material HDPE
Product Type Cap-style Hard Hat
Size 6.5 to 8
Ventilated Not Vented
Head Style Short Brim (Cap Style)