missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products Hard Hat Shade

Catalog No. p-7182062
Change view
Click to view available options
Color:
Blue
Orange
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19555123 White
19555125 Orange
19555126 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19555123 Supplier Pyramex Safety Products Supplier No. HPSHADE10
Only null left

Pyramex Safety™ Hard Hat Shade blocks sun from the face and neck on job sites lacking other forms of shade

  • Soft lightweight material
  • Hi-vis material for greater visibility
  • Fits on the outside of cap style and full brim hard hats

Specifications

Color White
For Use With (Equipment) Hard Hat