missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products Hard Hat Shade

Catalog No. 19555126
Change view
Click to view available options
Color:
Blue
Orange
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19555126 Blue
19555123 White
19555125 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19555126 Supplier Pyramex Safety Products Supplier No. HPSHADE60

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pyramex Safety™ Hard Hat Shade blocks sun from the face and neck on job sites lacking other forms of shade

  • Soft lightweight material
  • Hi-vis material for greater visibility
  • Fits on the outside of cap style and full brim hard hats

Specifications

Color Blue
For Use With (Equipment) Hard Hat