Learn More
3M™ Hard Hat, H-710R-UV
Description
- Protective hard hat with comfortable 4-point ratchet suspension and standard brow pad
- The hard hat comes in a variety of colours and features accessory slots for ear muff attachment and headgear, as well as a faceshield attachment
- The Uvicator sensor changes colour as the hard hat is exposed to UV light
- The sun’s UV rays can cause a hard hat shell to become brittle and compromise its ability to protect the wearer
- With the Uvicator sensor, the 3M™ Hard Hat should be replaced when the sun smart sensor changes from red to white
- The 4-point ratchet suspension adjusts easily to allow for a customized height and fit that best suits the individual wearer
- The ratchet system allows tightening or loosening without removing the hard hat, a hard hat fit for a variety of construction projects
Specifications
Specifications
| Color | Navy Blue |
| Product Type | Front Brim Hard Hat |
| Suspension Type | 4-point ratchet suspension with Uvicator |
| Ventilated | Not Vented |