Learn More
Hanna Instruments™ pH Electrode Cleaning Solution
Catalog No. 1160373
Encompass
Change view
Click to view available options
Capacity (Metric):
230 mL
Catalog No. Capacity (Metric)
1160373 230 mL
1 options
Catalog No. 1160373 Supplier Hanna Instruments™ Supplier No. HI 7061M
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly. Hanna Instruments™ pH Electrode Cleaning Solution dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
General purpose cleaning solution that can be used for a variety of applications.
  • Eliminates impurities and residues that are left on electrode surfaces when immersed in samples during measurement or when stored incorrectly.
  • Dissolves any mineral deposits or other coatings by soaking up to 20 minutes.
Specifications
Capacity (Metric) 230 mL
Filling Solution Type Aqueous
For Use With (Application) Removing impurities and residues from ph Electrodes