missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Checker™ pH Tester with 0.1 pH Resolution

Catalog No. 1330070
Encompass
Change view
Click to view available options
For Use With (Application):
pH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
1330070 pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1330070 Supplier Hanna Instruments™ Supplier No. HI98103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with virtually any electrode that shares same connector.

  • pH range from 0.00 to 14.00
  • Provides high accuracy with 0.1 pH resolution
  • Battery type 1.5V approximately 1000 hours of continous use
  • RH max 95%
  • With LCD and manual two point calibration
  • HI1271 pH electrode with screw type connector (included)

Specifications

For Use With (Application) pH
Accuracy ±0.2pH
Detection Range 0 to 14 pH