missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Presterilized
GREENER_CHOICE

Catalog No. 14222834
Change view
Click to view available options
Volume (Metric):
20 μL
50 μL
200 μL
1000 μL
Filtered:
Filtered
Non-filtered
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered
14222834 1000 μL Non-filtered
14222839 200 μL Non-filtered
14222852 1000 μL Filtered
14222855 200 μL Filtered
14222857 20 μL Filtered
14222860 50 μL Filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14222834 Supplier Axygen™ Supplier No. TT1000CHTRS

Clear tips for accurate sample delivery

Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.


  • 1000 μL Clear Tips
  • 96 Tip format
  • Clear Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Sterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head
  • Certifications

    Certified DNAse-, RNAse- and Pyrogen-safe

    Specifications

    Volume (Metric) 1000 μL
    Filtered Non-filtered
    Sterility Sterile
    Color Clear
    Autoclavable Autoclavable
    Certifications/Compliance Non-ANSI
    For Use With (Application) For accurate sample delivery
    Format Rack
    Tip Style Automation, Round
    Packaging Hanging tip rack
    Quantity 16 Racks/Cs.
    For Use With (Equipment) Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
    Material Polypropylene
    Show More Show Less