missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Clear Tips, Nonsterile
GREENER_CHOICE

Catalog No. 14222838
Change view
Click to view available options
Volume (Metric):
50 μL
200 μL
1000 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14222838 200 μL
14222848 50 μL
14222833 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14222838 Supplier Axygen™ Supplier No. TT200CHTR

Clear tips for accurate sample delivery

Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.


  • 1000 μL Clear Tips
  • 96 Tip format
  • Clear Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head
  • Certifications

    Certified DNAse-, RNAse- and Pyrogen-safe

    Specifications

    Volume (Metric) 200 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Autoclavable Autoclavable
    Certifications/Compliance Non-ANSI
    For Use With (Application) For accurate sample delivery
    Format Rack
    Tip Style Automation, Round
    Packaging Hanging tip rack
    Quantity 24 Racks/Cs.
    For Use With (Equipment) Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
    Material Polypropylene
    Show More Show Less