missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Hanger for Slotted Mass Set
For use with slotted mass set.
Supplier: Eisco™ PHZNW100H
Description
- Zinc die-casted with powder-coated
- Accuracy ±2%
Spécifications
| Powder-coated Hanger | |
| Zinc Die-cast | |
| 100 g | |
| Slotted Mass Set |