missing translation for 'onlineSavingsMsg'
Learn More

Moldex™ HandyStrap™ Series N95 Particulate Respirators

Catalog No. p-7237789
Change view
Click to view available options
Size:
Medium/Large
With Exhalation Valve:
No
Yes
Nuisance Odor Relief:
None
Organic Vapors
Color:
Black
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size With Exhalation Valve Nuisance Odor Relief Color
19039694B Medium/Large No None White
19037501 Medium/Large Yes None White
191600164 Medium/Large Yes None Black
19039695B Medium/Large Yes Organic Vapors White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19039694B Supplier Moldex™ Supplier No. 2600N95
Only null left

N95 disposable respirators feature a unique HandyStrap™ with buckle, allowing the mask to hang down around the user’s neck when not in use.

  • Cloth strap is cool and comfortable against the skin
  • HandyStrap™ with buckle, allows mask to hang down around the user's neck when not in use
  • Dura-Mesh™ provides bi-directional protection from dirt on the outside and moisture on the inside
  • Soft foam nose cushion for added comfort and no pressure points
  • Softspun™ lining for increased comfort and durability
  • Ventex™ exhalation valve lets the hot air out faster to enable easier breathing (all models EXCEPT 19-039-694A and -694B)
  • Special Ops™ models (19-160-0164 and 19-130-1352) are designed for military, armed forces, law enforcement and personal use
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1
  • N95 Plus Nuisance Organic Vapor Relief Applications (19-039-695B and 19-130-1352)

Specifications

Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2003, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Dura-Mesh
Product Type General Purpose Particulate Respirator
Size Medium/Large
With Exhalation Valve No
Filter Efficiency 95%
For Use With (Application) Grinding, sanding, sweeping, quarrying, machining and other dusty environments
Material Headstrap Cloth
Model 2600N95
Nose Piece Material Soft foam
Nuisance Odor Relief None
Quantity 15/Pk.
Strap Attachment Type HandyStrap™ with Buckle
FDA Cleared Surgical Mask No
Show More Show Less