Learn More
Research Products Corporation HandyFuge Plate Centrifuge, Accommodates Two 96-Well Plates
Catalog No. 50550337
Encompass
The HandyFuge from RPI provides a low cost and energy efficient method for spinning 96-well PCR plates, assay plates and 1.5-2.0ml micro-tubes. Ideal for use in RTPCR procedures and for maximizing sample recovery from micro-tubes. Because HandyFuge operates without electricity, it can be moved quickly to different locations and even be used in the field or in remote areas.
Accommodates two 96-well plates (skirted or non-skirted and up to 76 x 1.5-2.0ml micro-tubes). All it takes to operate is simply pushing the lever a few times and plates or tubes are ready for analysis. HandyFuge is designed for laboratory use and constructed from heavy duty components to provide years of dependable service.
Catalog No. 50550337 Supplier Research Products Corporation Supplier No. RPI 400752
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The HandyFuge from RPI provides a low cost and energy efficient method for spinning 96-well PCR plates, assay plates and 1.5-2.0ml micro-tubes. Ideal for use in RTPCR procedures and for maximizing sample recovery from micro-tubes. Because HandyFuge operates without electricity, it can be moved quickly to different locations and even be used in the field or in remote areas.
Accommodates two 96-well plates (skirted or non-skirted and up to 76 x 1.5-2.0ml micro-tubes). All it takes to operate is simply pushing the lever a few times and plates or tubes are ready for analysis. HandyFuge is designed for laboratory use and constructed from heavy duty components to provide years of dependable service.