missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hourglass International™ HandPRO™ Sable600™ Black Nitrile Gloves

Description
- Made from patented LOW DERMA technology nitrile to reduce your risk of glove-related allergic contact dermatitis
- Accelerator-free nitrile with no thiurams, carbamates or MBTs
- Chemotherapy drug permeation tested
- Fentanyl permeation tested (>240 minutes; 510(k) Granted by U.S. FDA)
- Excellent chemical resistance
- High tactile sensitivity
- Finger-textured for a secure grip
- Bold black color hides stains during use and makes light colored contaminants more visible
- Meets ASTM D6978 and USP 800 PPE requirement for use of chemotherapy gloves when handling hazardous drugs
- Meets FDA and EU requirements for food handling
- 300-count box maximizes space, reduces waste and minimizes cost
- Fits standard size glove box holder
- Powder-free
Specifications
Specifications
| Certifications/Compliance | ASTM D6319 Nitrile Exam Gloves for Medical Application; ASTM D6978 Chemotherapy Drugs and Fentanyl Citrate Tested; ASTM F1671 Viral Penetration Test |
| Certifications/Compliance (Europe) | EN 455 Nitrile Exam Gloves for Medical Application |
| Chemotherapeutic Rated | Yes |
| Chlorinated | Yes |
| Accelerator Free | Yes |
| Applications | Healthcare, Laboratory |
| AQL | 1.5 |
| Color | Black |
| Disposable | Single-use |
| Exterior Finish | Fingertip Textured |
| Show More |