missing translation for 'onlineSavingsMsg'
Learn More

Hourglass International™ HandPRO™ FreeStyle1100™ Nitrile Exam Gloves

Catalog No. 19423016
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19423016 2X-Small
19423006 X-Small
19423007 Small
19423008 Medium
19423009 Large
19423010 X-Large
15423050 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19423016 Supplier Hourglass International™ Supplier No. HG1106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nitrile exam gloves with low dermatitis potential are made from patented, accelerator-free LOW DERMA™ Technology nitrile to reduce the risk of developing type IV chemical allergies.

  • Made from patented LOW DERMA technology nitrile without accelerators
  • Manufactured without dithiocarbamates, thiurams or mercaptobenzothiazoles
  • Not made with natural rubber latex
  • Anti-slip super grip compound for wet and dry grip
  • Chemotherapy drug and Fentanyl Citrate permeation tested
  • Reduced risk of Type IV chemical allergies
  • Textured fingertips for enhanced tactile sensitivity
  • Powder free

Specifications

Certifications/Compliance ASTM D6319, ASTM D6978-05, ASTM F1671, FDA 21 CFR 177.2600, USP 800
Certifications/Compliance (Europe) EN455, European Commission Regulation (EU) No. 10/2011
Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free Yes
Applications Healthcare, Laboratory, Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 4.7 mil
Finger Thickness (Metric) 0.12 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.7 mil
Palm Thickness (Metric) 0.07 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 2X-Small
Sterility Non-sterile
Thickness (English) 4.7 mil
Thickness (Metric) 0.12 mm
FDA 510 (k) Yes
Model 1106
Series HandPRO™ Freestyle 1100™
Tensile Strength 14 MPa
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less