missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials HandE-Vac Handheld Aspirating System

Catalog No. p-7116141
Change view
Click to view available options
Model:
EV500
EV514
Type:
Handheld Aspirator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Type
10987035 EV514 Handheld Aspirator
10987042 EV500 Handheld Aspirator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10987035 Supplier Cole-Parmer™ Essentials Supplier No. 1305035

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Single channel adapter for disposable tip

A complete set of tools for aspiration—rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes with the Argos Technologies HandE-Vac Handheld Aspirator.

  • Use with Pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications
  • Pressure-sensitive button on the top of the unit controls the specific amount of vacuum appliedOperates with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems
  • Autoclavable

Specifications

Compatible Tips E-Vac Only
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Black
Disposable Reusable
Grip Type Ergonomic
Material Plastic
Model EV514
Number of Channels 1
Product Line HandE-Vac
Type Handheld Aspirator
Volume (Metric) 4 L
For Use With (Application) M-Vac or E-Vac aspiration systems
For Use With (Equipment) E-Vac™, M-Vac™ aspirating systems, or in-house vacuum source
Includes Single channel adapter for disposable tips
Show More Show Less