missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials HandE-Vac Handheld Aspirating System - Accessories

Catalog No. 10987036
Change view
Click to view available options
Model:
EV503
EV520
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10987036 EV520
10987034 EV503
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10987036 Supplier Cole-Parmer™ Essentials Supplier No. 1305037

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Set of tools for aspiration.

Operates with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems

Specifications

Compatible Tips 200 μL
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Black
Disposable Reusable
Grip Type Ergonomic
Material Plastic
Model EV520
Number of Channels 8
Product Line HandE-Vac
Type Tip Adapter
Volume (Metric) 4 L
For Use With (Application) HandE-Vac hand held aspirator
For Use With (Equipment) HandE-Vac hand held aspirator
Includes Ejector for disposable tips
Show More Show Less