missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials HandE-Vac Handheld Aspirating System - Accessories

Catalog No. 10987034
Change view
Click to view available options
Model:
EV503
EV520
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10987034 EV503
10987036 EV520
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10987034 Supplier Cole-Parmer™ Essentials Supplier No. 0439607

Set of tools for aspiration.

Operates with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems

Specifications

Compatible Tips E-Vac Only
Description 8-Channel stainless steel adapter
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Black
Disposable Reusable
Grip Type Ergonomic
Material Stainless Steel
Model EV503
Number of Channels 8
Product Line HandE-Vac
Type Tip Adapter
Volume (Metric) 4 L
For Use With (Application) HandE-Vac hand held aspirator
For Use With (Equipment) HandE-Vac hand held aspirator
Show More Show Less