Learn More
Thermo Scientific™ Quick Disconnect Drain Kit
Catalog No. 14511109
Change view
Click to view available options
Pump Type:
Drain Pump
Catalog No. Pump Type
14511109 Drain Pump
1 options
Catalog No. 14511109 Supplier Thermo Scientific™ Supplier No. 098246
Drain your water bath with a drain kit.
Drain your Thermo Scientific™ Precision™ water bath with a drain kit.
Drain Kit
  • Quickly drain shaking, circulating, or coliform baths with this drain kit
  • Includes: Hose, quick disconnect insert, quick disconnect valve
Specifications
Description Drain Kit
Pump Type Drain Pump
Type Quick Drain Kit