missing translation for 'onlineSavingsMsg'
Learn More

GOJO™ HAND MEDIC™ Professional Skin Conditioner

Catalog No. 296353303
Change view
Click to view available options
Volume (Metric):
147.87 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
296353303 147.87 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296353303 Supplier GOJO™ Supplier No. 815012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prevent dry, cracked skin by maintaining skin moisture levels. Professional skin conditioner is specially formulated for industrial environments.

  • Fragrance-free skin conditioner
  • Specially formulated for industrial environments
  • Helps prevent dry, cracked skin by maintaining skin's moisture level
  • Absorbs quickly with no greasy after-feel
  • Silicone and fragrance free
  • Dermatologist tested
  • Latex glove compatible
  • NSF E4 Registered

Specifications

Product Type Skin Conditioner
Color White
Form Lotion
For Use With (Application) Helps prevent dry, cracked skin by maintaining skin moisture level
Quantity 12/Pk.
Size 5 oz.
Volume (Metric) 147.87 mL