missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Hand Lens, Plastic Frame

Durable design for repeated use.

Supplier:  United Scientific™ MFP050

 View more versions of this product

Catalog No. S16011


Only null left
Explore available promotions

Description

Description

  • Glass lens, molded into high-quality plastic frame.
  • Available in various diameters
  • Provides 2.5x magnification
Specifications

Specifications

50 mm
Glass, Plastic
2.5x
Magnifier
2 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Warning: Cancer and reproductive harm