Learn More
MilliporeSigma™ Supelco™ Hand Crimper, Adjustable
Catalog No. p-7194175
Change view
Click to view available options
For Use With (Equipment):
11 mm seals
20 mm seals
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
111009381 11 mm seals
111009382 20 mm seals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111009381 Supplier MilliporeSigma™ Supelco™ Supplier No. 22313U
Only null left
The adjustable hand crimper is ideal if you use varying thickness of septa in your analysis. This product has the ability to be adjusted for changes in septa thickness. This product is very useful in SPME applications recommending the use of a thinner septa (1.0-1.5 mm thickness).
Specifications
Description Adjustable Hand Crimper
No. per Pack 1/Pk.
For Use With (Equipment) 11 mm seals