Learn More
Hamilton™ VDRL Antigen Dispenser
Description
Designed to be used with the PB600 to dispense liquid up to 50 times without refilling. Needle is bent at 90 degrees which allows easy syringe reading and filling.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Borosilicate Glass |
| Product Type | VDRL Antigen Dispenser |
| Needle Point Number | No Needle Included |
| Volume (Metric) | 833 μL |
| For Use With (Equipment) | PB600 |
| Connection Type | Luer |
| Sterility | Sterile |