Learn More
Hamilton™ Needle Cleaning Concentrate
A biodegradable, non-phosphate cleaning concentrate that is suitable for many common sample types.
$109.50 - $153.30
Specifications
| Product Type | Cleaning Concentrate |
|---|---|
| For Use With (Equipment) | Syringe and Needle Cleaning Kit |
Description
- Non-alkaline, non-phosphate and non-detergent based
- 70mL or 500mL bottle
- For syringes and needles
Specifications
| Cleaning Concentrate | |
| Syringe and Needle Cleaning Kit |