Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
Supplier: Hamilton™ 5900045
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Chemical Identifiers
| Microlab™ 600 dispensers |
Specifications
| 1005 BFP | |
| 5 mL | |
| Microlab™ 600 dispensers |