Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessories - (BFP) Syringes
For use with MICROLAB 600 Diluter/Dispenser
Supplier:  Hamilton™ 5900035
 View more versions of this product
Catalog No. 14816275
Only null left
Description
Description
Syringe has a conical plunger tip that extends through the threaded termination and into the valve. This unique design expels the air from the syringe and valve decreasing the number of priming cycles required.
Specifications
Chemical Identifiers
Microlab™ 600 dispensers
Specifications
1001 BFP
1 mL
Microlab™ 600 dispensers
Product Suggestions
Product Suggestions