Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Upgrade Kit
Catalog No. 14816289
Change view
Click to view available options
For Use With (Equipment):
Microlab 600 Diluter/Dispenser
Catalog No. For Use With (Equipment)
14816289 Microlab 600 Diluter/Dispenser
1 options
Catalog No. 14816289 Supplier Hamilton™ Supplier No. 6150003
Control the Microlab 600 Pump without a controller by using your computer.
Each program is able to perform simple pump movements and communications. The list below is the full extent of the features included in the example programs.
  • Syringe size selection
  • Initialization
  • Priming
  • Valve positioning
  • Single step syringe movements
  • Trigger probe functionality
The example programs will not be able to program multi-step custom applications, automatically refill on empty, select preset valve configurations, create air gaps or rinse steps, etc. If any of these functionalities are required for your application it is highly recommended you have a skilled programmer available to create these features in your own custom program. If you are unsure of the features required for your application, please contact Hamilton tech support at laboratory@hamiltoncompany.com to discuss the best possible options. Hamilton does not currently provide programming support and will not be able to assist with any further matters related PC controlled Microlab 600 usage
Specifications
Type Custom Programmer Kit
For Use With (Equipment) Microlab 600 Diluter/Dispenser
Includes Programmer Software CD with Programmer Manual, Application Programming Interface, and Example LabView, C#, and VB Programs