missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ HPLC Reverse Phase Bulk Packing Resin, Sold by MilliporeSigma™ Supelco™

Catalog No. 111012156
Change view
Click to view available options
100 Å
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111012156 100 Å
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012156 Supplier Merck Emd Millipore Supplier No. HAM795811EA
Only null left

PRP-1, Particle Size: 12-20 μm

  • PSDVB Packing
  • Maximum Pressure: 5000 psi

Specifications

Particle Size 12 to 20 μm
Pore Size 100 Å
Quantity 1 Ea.
Product Line Hamilton
Description PSDVB Packing
Type Bulk Resin

Hamilton is a registered trademark of Hamilton Co.