Learn More
Hamilton™ Fill Tubing
Supplier: Hamilton™ HAM240000
Specifications
| PTFE | |
| 25.59 in. | |
| Fill Tubing | |
| ML500 valves with M6 threads, used with volumes greater than 1mL syringes |
| 12 G | |
| 65 cm | |
| 2042 μL |