missing translation for 'onlineSavingsMsg'
Learn More

Halosulfuron-methyl 98.0+%, TCI America™

Catalog No. H13741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13741G 1 g
H13745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13741G Supplier TCI America Supplier No. H13741G

Chemical Identifiers

CAS 100784-20-1
Molecular Formula C13H15ClN6O7S
Molecular Weight (g/mol) 434.808
MDL Number MFCD01631160
InChI Key FMGZEUWROYGLAY-UHFFFAOYSA-N
Synonym Methyl 3-Chloro-5-[[3-(4,6-dimethoxypyrimidin-2-yl)ureido]sulfonyl]-1-methyl-1H-pyrazole-4-carboxylate, 3-Chloro-5-[[3-(4,6-dimethoxypyrimidin-2-yl)ureido]sulfonyl]-1-methyl-1H-pyrazole-4-carboxylic Acid Methyl Ester
PubChem CID 91763
ChEBI CHEBI:81750
IUPAC Name methyl 3-chloro-5-[(4,6-dimethoxypyrimidin-2-yl)carbamoylsulfamoyl]-1-methylpyrazole-4-carboxylate
SMILES CN1C(=C(C(=N1)Cl)C(=O)OC)S(=O)(=O)NC(=O)NC2=NC(=CC(=N2)OC)OC

Specifications

Melting Point 174°C
Quantity 1 g
UN Number 3077
Formula Weight 434.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Halosulfuron-methyl
RTECSNumber UQ6392606
TSCA No