missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol

Catalog No. AAJ6168806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6168806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6168806 Supplier Thermo Scientific Chemicals Supplier No. J6168806

CAS: 52-86-8 | C21H23ClFNO2 | 375.87 g/mol

An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

An antagonist for D2, D3, and D4 dopamine receptors that induces apoptosis of neurons in the striatum of rats.

Specifications

CAS 52-86-8
Color White
Molecular Formula C21H23ClFNO2
Quantity 5 g
MDL Number MFCD00051423
UN Number UN2811
Merck Index 14,4598
Synonym haloperidol, haldol, aloperidin, serenace, aloperidol, brotopon, eukystol, einalon s, aloperidolo, galoperidol
InChI Key LNEPOXFFQSENCJ-UHFFFAOYSA-N
SMILES OC1(CCN(CCCC(=O)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C=C1
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
Molecular Weight (g/mol) 375.87
PubChem CID 3559
ChEBI CHEBI:5613
Formula Weight 375.86
Physical Form Powder
Chemical Name or Material Haloperidol
Show More Show Less
Hazard Category H301-H315-H317-H319-H335-H360
Hazard Statement GHS H Statement
H301-H360-H315-H319-H317-H335
Toxic if swallowed.
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-155-6
RTECSNumber EU1575000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only