missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol 98.0+%, TCI America™

Catalog No. p-7130803
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09125G 5 g
H091225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09125G Supplier TCI America Supplier No. H09125G

Chemical Identifiers

CAS 52-86-8
Molecular Formula C21H23ClFNO2
Molecular Weight (g/mol) 375.87
MDL Number MFCD00051423
InChI Key LNEPOXFFQSENCJ-UHFFFAOYSA-N
Synonym haloperidol, haldol, aloperidin, serenace, aloperidol, brotopon, eukystol, einalon s, aloperidolo, galoperidol
PubChem CID 3559
ChEBI CHEBI:5613
IUPAC Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-1-(4-fluorophenyl)butan-1-one
SMILES OC1(CCN(CCCC(=O)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 152°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 375.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Haloperidol
EINECSNumber (9)-1138
RTECSNumber EU1575000
TSCA No