missing translation for 'onlineSavingsMsg'
Learn More

Bulbtronics™ Halogen Lamps: Mercury Arc

Catalog No. 14395374
Encompass
Change view
Click to view available options
Wattage:
50 W
100 W
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wattage
14395374 100 W
14395371 50 W
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Catalog No. 14395374 Supplier Bulbtronics™ Supplier No. 294
Only null left

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Specifications

Base Style Sleeve
Light Source Mercury Arc
Lighting Life 200 hr.
Voltage 120 V
Wattage 100 W
Color Clear
Diameter (English) 0.39 in.
Diameter (Metric) 0.99 cm
For Use With (Equipment) Microscope
Length (English) 3.54 in.
Length (Metric) 9 cm
Lumen 17,0000 lm
Shape T4
Product Type Mercury Arc Lamp
Show More Show Less