missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulbtronics™ Halogen Lamps: Mercury Arc
Supplier: Bulbtronics™ 268
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications
| SFA6-2 | |
| 100 hr. | |
| Clear | |
| 0.83 cm | |
| 2.08 in. | |
| 5.3 cm | |
| 2000 lm |
| Mercury Arc | |
| 50 W | |
| 0.33 in. | |
| Microscope | |
| 0.08 in. | |
| 0.22 cm | |
| Mercury Arc Lamp |