Learn More
Impacto™ Half-Finger Anti-Impact Mesh Carpal Tunnel Gloves
Catalog No. 19400093
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19400093 Small
19400094 Medium
19400095 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19400093 Supplier Impacto™ Supplier No. ST861020
Gloves feature a unique channel that takes the pressure off of the median nerve, which alleviates symptoms of repetitive strain injuries such as carpal tunnel syndrome. VEP padding in the palm dissipates shock, while the half-finger design maximizes tactile feedback.
  • Half-finger enhances tactile feedback
  • Durable cowhide leather palm with VEP padding dissipates shock
  • Air mesh back increases ventilation
  • Gusseted fingers for comfort
Specifications
Back Material Nylon Mesh
Color Black
Material Cowhide Leather
Size Small
Finger Style Half
Glove Cuff Style Hook and Loop
Series ST8610