missing translation for 'onlineSavingsMsg'
Learn More

Hafnium wire, 0.5mm (0.02in) dia, 99.95% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Catalog No. p-7058667
Change view
Click to view available options
Quantity:
25 cm
100 cm
500 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42982BU 25 cm
AA42982BY 100 cm
AA42982CC 500 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42982BU Supplier Thermo Scientific Chemicals Supplier No. 042982BU
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]

Specifications

Quantity 25 cm
Odor Odorless
Weight ≈2.6g/m
Form Wire
Percent Purity 99.95%
Assay (metals basis excluding Zr), Zr nominal 3%
Chemical Name or Material Hafnium wire, 0.5mm (0.02 in.) dia.
EINECSNumber 231-166-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only