missing translation for 'onlineSavingsMsg'
Learn More

Hafnium telluride, 99.5% (metals basis)

CAS: 39082-23-0 | HfTe2 | 433.69 g/mol

$298.34 - $1454.62

Chemical Identifiers

CAS 39082-23-0
Molecular Formula HfTe2
Molecular Weight (g/mol) 433.69
MDL Number MFCD00049947
InChI Key BCMXGUYIVFZMCT-UHFFFAOYSA-N
Synonym hafnium telluride, ditellanylidenehafnium
PubChem CID 90471877
IUPAC Name bis(tellanylidene)hafnium
SMILES [Te]=[Hf]=[Te]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3922303
View Documents
Thermo Scientific Chemicals
03922303
1 g
Each for $298.34
Only null left
 
AA3922306
View Documents
Thermo Scientific Chemicals
03922306
5 g
Each for $1,454.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39082-23-0
433.69
BCMXGUYIVFZMCT-UHFFFAOYSA-N
90471877
[Te]=[Hf]=[Te]
HfTe2
MFCD00049947
hafnium telluride, ditellanylidenehafnium
bis(tellanylidene)hafnium

Specifications

39082-23-0
1 g
MFCD00049947
BCMXGUYIVFZMCT-UHFFFAOYSA-N
bis(tellanylidene)hafnium
90471877
99.5%
(metals basis)
Powder
HfTe2
hafnium telluride, ditellanylidenehafnium
[Te]=[Hf]=[Te]
433.69
433.69
Hafnium telluride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

EINECSNumber : 254-280-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only