missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) fluoride, 99.9% (metals basis), Thermo Scientific Chemicals

$925.01 - $925.01

Chemical Identifiers

CAS 13709-52-9
Molecular Formula HfF4
MDL Number MFCD00016127
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4506114
View Documents
Thermo Scientific Chemicals
04506114
25 g
Each for $925.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13709-52-9
HfF4
MFCD00016127

Specifications

White
Powder
HfF4
UN3290
99.9%
Hafnium (IV) fluoride
13709-52-9
25 g
MFCD00016127
254.48
Moisture sensitive
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 237-258-0

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only