missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%

Catalog No. AA4561236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4561236 500 g
AA4561214 25 g
AA4561222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4561236 Supplier Thermo Scientific Chemicals Supplier No. 04561236

CAS: 13499-05-3 | Cl4Hf | 320.29 g/mol

Hafnium(IV) chloride is used as a precursor for catalyst involved in the Ziegler-Natta polymerization of propylene. It is an effective Lewis acid for various applications in organic synthesis viz alkylation of ferrocene. Further, it increases the rate and control of 1,3-dipolar cycloadditions. It is also used to prepare hafnium dioxide and hafnium silicate, which is used as high-k-dielectrics in the production of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13499-05-3
Molecular Formula Cl4Hf
Molecular Weight (g/mol) 320.29
MDL Number MFCD00011034
InChI Key PDPJQWYGJJBYLF-UHFFFAOYSA-J
Synonym hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis
PubChem CID 37715
IUPAC Name tetrachlorohafnium
SMILES Cl[Hf](Cl)(Cl)Cl

Specifications

Melting Point 320° (sublimation)
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Soluble in methanol and acetone.
Formula Weight 320.3
Odor Irritating
Sensitivity Moisture sensitive
Chemical Name or Material Hafnium(IV) chloride
Hazard Category H314
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 236-826-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only