missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) chloride, 98+% (metals basis excluding Zr), Zr <2.7%

Catalog No. AA1183488
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1183488 2.5 g
AA1183414 25 g
AA1183422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1183488 Supplier Thermo Scientific Chemicals Supplier No. 01183488

CAS: 13499-05-3 | Cl4Hf | 320.29 g/mol

Hafnium chloride tetrahydrofuran complex is an efficient catalyst for the Diels-Alder cycloadditions, Ziegler-Natta polymerization of alkenes. It is used as control rods in nuclear reactors and in manufacture of light bulb filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13499-05-3
Molecular Formula Cl4Hf
Molecular Weight (g/mol) 320.29
MDL Number MFCD00011034
InChI Key PDPJQWYGJJBYLF-UHFFFAOYSA-J
Synonym hafnium tetrachloride, hafnium iv chloride, hafnium chloride hfcl4 , t-4, hfcl4, hafnium iv chloride,hafnium tetrachloride, hafnium iv chloride, sublimed grade, hafnium iv chloride, purum at, hafnium iv chloride, purified by sublimation trace metals basis
PubChem CID 37715
IUPAC Name tetrachlorohafnium
SMILES Cl[Hf](Cl)(Cl)Cl

Specifications

Melting Point 320° (sublimation)
Color White
Physical Form Powder
Quantity 2.5 g
Assay Percent Range >98% (Metals basis excluding Zr)
Solubility Information Soluble in water,methanol and acetone.
Formula Weight 320.3
Odor Irritating
Sensitivity Moisture sensitive
Chemical Name or Material Hafnium(IV) chloride
Hazard Category H314
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 236-826-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only