Learn More
HTL-Strefa DropSafe Haemolance Plus™ Safety Lance
Safety lancets designed for safety, comfort, and simplicity. Fully hidden needle. Silicone-coated, ultra-sharp needle. Easy to handle, intuitive activation. Designed for convenience and ideal blood sample collection.
$52.93
Specifications
| Product Type | Lancet |
|---|---|
| Disposable | Single-use |
| Quantity | 100/Box |
| Sterility | Sterile |
| Catalog Number | Mfr. No. | Color | Depth (Metric) Needle | Needle Gauge | Flow Type | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Depth (Metric) Needle | Needle Gauge | Flow Type | Price | Quantity & Availability | ||
|
22192014
|
HTL-Strefa
7372 |
Green | 1.8 mm | 21 G | Normal |
Pack of 100 Each for $52.93
Case of 20 Pack for $1,058.60
|
|
||
Description
- Button Activated Safety lancets designed for safety, comfort, and simplicity.
- Safety
- All needles are irradiation-sterilized and protected with a protective cap. Fully enclosed needle housing ensures that the needle is hidden before and after use to prevent sharps injuries. Auto-Retraction mechanism prevents reuse of the device.
- Comfort
- Silicone-coated, ultra-sharp needles, ideally positioned for skin penetration. Various, color-coded product versions to meet specific blood sample applications. Wide range of designs providing a simple, comfortable, and secure puncture.
- Simplicity
- Easy to handle, intuitive activation with no pre-loading for both push button and pressure activations. Easy to grasp protective cap for easy removal. Simply twist and pull to remove the protective cap. Designed for convenient finger positioning and ideal blood sample collection.
- Ready to Use
- The lancet is activated by first removing the safety cap, pressing the lancet against the fingertip, then depressing the push-button.
- In use
- During activation, the wings on the needle support break, which releases the needle.
- After use
- After activation, the needle retracts back into the device. Immediately dispose of into an appropriate sharps disposal
Specifications
| Lancet | |
| Single-use | |
| 100/Box | |
| Sterile |