Learn More
Alkali Scientific H3022 Incu-Shaker 20LR Stackab
Catalog No. 503029602
Encompass
Stainless steel chamber provides premier temperature uniformity Proven reliable maintenance-free Incu-Shakermotor drive system Bright intuitive touch-screen controls Instantly change flask clamps with MAGic Clampaccessories Included pull-out adjustable shelf for static incubations The new Alkali Incu-Shaker20LR (H3022) introduce a high-throughput space saving shaking incubator system to the Incu-Shaker family Now with stacking capability of up to 2 units the Incu-Shaker20 series are highly space-efficient without compromising on reliability or temperature performance Thoroughly tested for years of reliable performance the same proven brushless induction motor drive is featured as in other Incu-Shaker instruments The high grade stainless-steel internal chambers provide temperature accuracy within 0 1C and industry-leading uniformity within the chamber of 0 5C The Incu-Shaker20L features temperature control from 5C above ambient temperatures up to 60C The 20LR features a built-in po
Catalog No. 503029602 Supplier Alkali Scientific Supplier No. RSH3022
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stainless steel chamber provides premier temperature uniformity Proven reliable maintenance-free Incu-Shakermotor drive system Bright intuitive touch-screen controls Instantly change flask clamps with MAGic Clampaccessories Included pull-out adjustable shelf for static incubations The new Alkali Incu-Shaker20LR (H3022) introduce a high-throughput space saving shaking incubator system to the Incu-Shaker family Now with stacking capability of up to 2 units the Incu-Shaker20 series are highly space-efficient without compromising on reliability or temperature performance Thoroughly tested for years of reliable performance the same proven brushless induction motor drive is featured as in other Incu-Shaker instruments The high grade stainless-steel internal chambers provide temperature accuracy within 0 1C and industry-leading uniformity within the chamber of 0 5C The Incu-Shaker20L features temperature control from 5C above ambient temperatures up to 60C The 20LR features a built-in po