Learn More
Abcam Inc H-Met-AMC, Calpain substrate, 100MG
Catalog No. 502705158
Encompass
MW 420.4 g/mol, Purity >99%. Calpain substrate. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 502705158 Supplier Abcam Inc Supplier No. AB145498100MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MW 420.4 g/mol, Purity >99%. Calpain substrate. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement