missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Spin more with less! High capacity and flexibility—all in a small footprint.
Supplier: Thermo Scientific™ 75003300
Description
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
- Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Chemical Identifiers
| Set of 2 |
Specifications
| Cross | |
| Swinging Bucket Rotor | |
| 4297 | |
| Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series | |
| Biocontainment Certified by CAMR™, HPA, Porton Down, UK | |
| Autoclavable |
| Cast/Machined Steel with Slide Coating | |
| See Individual Adapter Capacities | |
| Rmax 174 mm | |
| 90° | |
| Yes, when used with biocontainment lids | |
| Set of 2 |